Anti-ALB Mouse Monoclonal Antibody [clone: 3A10]

Proveedor: OriGene
CF507291
ORIGCF507291EA 764 EUR
ORIGCF507291
Anti-ALB Mouse Monoclonal Antibody [clone: 3A10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ALB
Clonality: Monoclonal
Clone: 3A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR