Anti-DDX56 Mouse Monoclonal Antibody [clone: 1G6]

Proveedor: OriGene
CF802941
ORIGCF802941EA 764 EUR
ORIGCF802941
Anti-DDX56 Mouse Monoclonal Antibody [clone: 1G6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DDX56
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR