Anti-CSH1 Mouse Monoclonal Antibody [clone: 4D6]

Proveedor: OriGene
CF506816
ORIGCF506816EA 764 EUR
ORIGCF506816
Anti-CSH1 Mouse Monoclonal Antibody [clone: 4D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: CSH1
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR