Anti-BID Mouse Monoclonal Antibody [clone: 4A1]

Proveedor: OriGene
CF506762
ORIGCF506762EA 786 EUR
ORIGCF506762
Anti-BID Mouse Monoclonal Antibody [clone: 4A1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: BID
Clonality: Monoclonal
Clone: 4A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR