Anti-CD44 Mouse Monoclonal Antibody [clone: 5C4]

Proveedor: OriGene
CF506740
ORIGCF506740EA 764 EUR
ORIGCF506740
Anti-CD44 Mouse Monoclonal Antibody [clone: 5C4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:400-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 5C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR