Anti-CD44 Mouse Monoclonal Antibody [clone: 5D5]

Proveedor: OriGene
CF506739
ORIGCF506739EA 764 EUR
ORIGCF506739
Anti-CD44 Mouse Monoclonal Antibody [clone: 5D5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:400-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 5D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR