Anti-ACY1 Mouse Monoclonal Antibody [clone: 1B12]

Proveedor: OriGene
CF503227
ORIGCF503227EA 764 EUR
ORIGCF503227
Anti-ACY1 Mouse Monoclonal Antibody [clone: 1B12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ACY1
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR