Anti-ACY1 Mouse Monoclonal Antibody [clone: 1A12]

Proveedor: OriGene
CF503226
ORIGCF503226EA 764 EUR
ORIGCF503226
Anti-ACY1 Mouse Monoclonal Antibody [clone: 1A12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ACY1
Clonality: Monoclonal
Clone: 1A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR