Anti-ACY1 Mouse Monoclonal Antibody [clone: 2F1]

Proveedor: OriGene
CF503213
ORIGCF503213EA 764 EUR
ORIGCF503213
Anti-ACY1 Mouse Monoclonal Antibody [clone: 2F1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ACY1
Clonality: Monoclonal
Clone: 2F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR