Anti-PHF21B Mouse Monoclonal Antibody [clone: 4C5]

Proveedor: OriGene
CF503205
ORIGCF503205EA 764 EUR
ORIGCF503205
Anti-PHF21B Mouse Monoclonal Antibody [clone: 4C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PHF21B
Clonality: Monoclonal
Clone: 4C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR