Anti-PTPN7 Mouse Monoclonal Antibody [clone: 3H8]

Proveedor: OriGene
CF503662
ORIGCF503662EA 764 EUR
ORIGCF503662
Anti-PTPN7 Mouse Monoclonal Antibody [clone: 3H8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PTPN7
Clonality: Monoclonal
Clone: 3H8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR