Anti-CYP2J2 Mouse Monoclonal Antibody [clone: 3B4]

Proveedor: OriGene
CF503635
ORIGCF503635EA 764 EUR
ORIGCF503635
Anti-CYP2J2 Mouse Monoclonal Antibody [clone: 3B4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CYP2J2
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR