Anti-KIAA0495 Mouse Monoclonal Antibody [clone: 2B9]

Proveedor: OriGene
CF503633
ORIGCF503633EA 786 EUR
ORIGCF503633
Anti-KIAA0495 Mouse Monoclonal Antibody [clone: 2B9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: KIAA0495
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR