Anti-CA12 Mouse Monoclonal Antibody [clone: 3F1]

Proveedor: OriGene
ORIGCF505598EA 764 EUR
ORIGCF505598
Anti-CA12 Mouse Monoclonal Antibody [clone: 3F1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:50

Type: Primary
Antigen: CA12
Clonality: Monoclonal
Clone: 3F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR