Anti-RHOJ Mouse Monoclonal Antibody [clone: 5F5]

Proveedor: OriGene
CF505593
ORIGCF505593EA 764 EUR
ORIGCF505593
Anti-RHOJ Mouse Monoclonal Antibody [clone: 5F5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: RHOJ
Clonality: Monoclonal
Clone: 5F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR