Anti-GBAS Mouse Monoclonal Antibody [clone: 1D6]

Proveedor: OriGene
CF505585
ORIGCF505585EA 764 EUR
ORIGCF505585
Anti-GBAS Mouse Monoclonal Antibody [clone: 1D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GBAS
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR