Anti-CA12 Mouse Monoclonal Antibody [clone: 4D1]

Proveedor: OriGene
CF505569
ORIGCF505569EA 764 EUR
ORIGCF505569
Anti-CA12 Mouse Monoclonal Antibody [clone: 4D1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CA12
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR