Anti-GLB1 Mouse Monoclonal Antibody [clone: 2F6]

Proveedor: OriGene
CF505568
ORIGCF505568EA 764 EUR
ORIGCF505568
Anti-GLB1 Mouse Monoclonal Antibody [clone: 2F6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: GLB1
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR