Anti-TAPBPL Mouse Monoclonal Antibody [clone: 5E5]

Proveedor: OriGene
CF505554
ORIGCF505554EA 764 EUR
ORIGCF505554
Anti-TAPBPL Mouse Monoclonal Antibody [clone: 5E5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TAPBPL
Clonality: Monoclonal
Clone: 5E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR