Anti-ALDH3A2 Mouse Monoclonal Antibody [clone: 1B11]

Proveedor: OriGene
CF503196
ORIGCF503196EA 764 EUR
ORIGCF503196
Anti-ALDH3A2 Mouse Monoclonal Antibody [clone: 1B11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ALDH3A2
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR