Anti-ERCC4 Mouse Monoclonal Antibody [clone: 3H7]

Proveedor: OriGene
CF503163
ORIGCF503163EA 764 EUR
ORIGCF503163
Anti-ERCC4 Mouse Monoclonal Antibody [clone: 3H7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC4
Clonality: Monoclonal
Clone: 3H7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR