Anti-SERPINB6 Mouse Monoclonal Antibody [clone: 3H3]

Proveedor: OriGene
CF504355
ORIGCF504355EA 764 EUR
ORIGCF504355
Anti-SERPINB6 Mouse Monoclonal Antibody [clone: 3H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINB6
Clonality: Monoclonal
Clone: 3H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR