Anti-COPS5 Mouse Monoclonal Antibody [clone: 2B12]

Proveedor: OriGene
CF504344
ORIGCF504344EA 764 EUR
ORIGCF504344
Anti-COPS5 Mouse Monoclonal Antibody [clone: 2B12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: COPS5
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR