Anti-COG6 Mouse Monoclonal Antibody [clone: 2D8]

Proveedor: OriGene
CF505531
ORIGCF505531EA 764 EUR
ORIGCF505531
Anti-COG6 Mouse Monoclonal Antibody [clone: 2D8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: COG6
Clonality: Monoclonal
Clone: 2D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR