Anti-TRMT2A Mouse Monoclonal Antibody [clone: 3B7]

Proveedor: OriGene
CF505523
ORIGCF505523EA 786 EUR
ORIGCF505523
Anti-TRMT2A Mouse Monoclonal Antibody [clone: 3B7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TRMT2A
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR