Anti-BTLA Mouse Monoclonal Antibody [clone: 1E5]

Proveedor: OriGene
CF505519
ORIGCF505519EA 764 EUR
ORIGCF505519
Anti-BTLA Mouse Monoclonal Antibody [clone: 1E5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: BTLA
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR