Anti-TRIM44 Mouse Monoclonal Antibody [clone: 6B11]

Proveedor: OriGene
CF505511
ORIGCF505511EA 764 EUR
ORIGCF505511
Anti-TRIM44 Mouse Monoclonal Antibody [clone: 6B11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TRIM44
Clonality: Monoclonal
Clone: 6B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR