Anti-LCMT1 Mouse Monoclonal Antibody [clone: 4A5]

Proveedor: OriGene
CF503138
ORIGCF503138EA 764 EUR
ORIGCF503138
Anti-LCMT1 Mouse Monoclonal Antibody [clone: 4A5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LCMT1
Clonality: Monoclonal
Clone: 4A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR