Anti-CXorf26 Mouse Monoclonal Antibody [clone: 5A11]

Proveedor: OriGene
CF503131
ORIGCF503131EA 786 EUR
ORIGCF503131
Anti-CXorf26 Mouse Monoclonal Antibody [clone: 5A11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50-150

Type: Primary
Antigen: CXorf26
Clonality: Monoclonal
Clone: 5A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR