Anti-CST2 Mouse Monoclonal Antibody [clone: 5G8]

Proveedor: OriGene
CF504340
ORIGCF504340EA 786 EUR
ORIGCF504340
Anti-CST2 Mouse Monoclonal Antibody [clone: 5G8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CST2
Clonality: Monoclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR