Anti-DTNB Mouse Monoclonal Antibody [clone: 3H8]

Proveedor: OriGene
CF504332
ORIGCF504332EA 764 EUR
ORIGCF504332
Anti-DTNB Mouse Monoclonal Antibody [clone: 3H8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DTNB
Clonality: Monoclonal
Clone: 3H8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR