Anti-PPP1R15A Mouse Monoclonal Antibody [clone: 2B11]

Proveedor: OriGene
CF504309
ORIGCF504309EA 786 EUR
ORIGCF504309
Anti-PPP1R15A Mouse Monoclonal Antibody [clone: 2B11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPP1R15A
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR