Anti-ERBB2 Mouse Monoclonal Antibody [clone: 12G4]

Proveedor: OriGene
CF503527
ORIGCF503527EA 764 EUR
ORIGCF503527
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 12G4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 12G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR