Anti-MGLL Mouse Monoclonal Antibody [clone: 1B1]

Proveedor: OriGene
CF503068
ORIGCF503068EA 764 EUR
ORIGCF503068
Anti-MGLL Mouse Monoclonal Antibody [clone: 1B1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MGLL
Clonality: Monoclonal
Clone: 1B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR