Anti-MGLL Mouse Monoclonal Antibody [clone: 4D1]

Proveedor: OriGene
CF503067
ORIGCF503067EA 764 EUR
ORIGCF503067
Anti-MGLL Mouse Monoclonal Antibody [clone: 4D1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MGLL
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR