Anti-EPN2 Mouse Monoclonal Antibody [clone: 1C2]

Proveedor: OriGene
CF504297
ORIGCF504297EA 786 EUR
ORIGCF504297
Anti-EPN2 Mouse Monoclonal Antibody [clone: 1C2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EPN2
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR