Anti-DTYMK Mouse Monoclonal Antibody [clone: 1E12]

Proveedor: OriGene
CF503490
ORIGCF503490EA 764 EUR
ORIGCF503490
Anti-DTYMK Mouse Monoclonal Antibody [clone: 1E12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DTYMK
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR