Anti-KIF25 Mouse Monoclonal Antibody [clone: 1H10]

Proveedor: OriGene
CF505431
ORIGCF505431EA 786 EUR
ORIGCF505431
Anti-KIF25 Mouse Monoclonal Antibody [clone: 1H10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: KIF25
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR