Anti-ERCC4 Mouse Monoclonal Antibody [clone: 2D4]

Proveedor: OriGene
ORIGCF503063EA 764 EUR
ORIGCF503063
Anti-ERCC4 Mouse Monoclonal Antibody [clone: 2D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC4
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR