Anti-EIF4E Mouse Monoclonal Antibody [clone: 9H1]

Proveedor: OriGene
CF503029
ORIGCF503029EA 764 EUR
ORIGCF503029
Anti-EIF4E Mouse Monoclonal Antibody [clone: 9H1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: EIF4E
Clonality: Monoclonal
Clone: 9H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR