Anti-AP2M1 Mouse Monoclonal Antibody [clone: 2F11]

Proveedor: OriGene
CF503021
ORIGCF503021EA 764 EUR
ORIGCF503021
Anti-AP2M1 Mouse Monoclonal Antibody [clone: 2F11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: AP2M1
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR