Anti-LGALS3BP Mouse Monoclonal Antibody [clone: 6B7]

Proveedor: OriGene
CF503453
ORIGCF503453EA 764 EUR
ORIGCF503453
Anti-LGALS3BP Mouse Monoclonal Antibody [clone: 6B7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LGALS3BP
Clonality: Monoclonal
Clone: 6B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR