Anti-PMVK Mouse Monoclonal Antibody [clone: 2D1]

Proveedor: OriGene
CF503448
ORIGCF503448EA 764 EUR
ORIGCF503448
Anti-PMVK Mouse Monoclonal Antibody [clone: 2D1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: PMVK
Clonality: Monoclonal
Clone: 2D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR