Anti-CYP17A1 Mouse Monoclonal Antibody [clone: 5G10]

Proveedor: OriGene
CF503415
ORIGCF503415EA 764 EUR
ORIGCF503415
Anti-CYP17A1 Mouse Monoclonal Antibody [clone: 5G10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CYP17A1
Clonality: Monoclonal
Clone: 5G10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR