Anti-ARMC1 Mouse Monoclonal Antibody [clone: 1D10]

Proveedor: OriGene
CF505405
ORIGCF505405EA 786 EUR
ORIGCF505405
Anti-ARMC1 Mouse Monoclonal Antibody [clone: 1D10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ARMC1
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR