Anti-TSC22D3 Mouse Monoclonal Antibody [clone: 1H3]

Proveedor: OriGene
ORIGCF505355EA 764 EUR
ORIGCF505355
Anti-TSC22D3 Mouse Monoclonal Antibody [clone: 1H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TSC22D3
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR