Anti-PRKAR1B Mouse Monoclonal Antibody [clone: 12E5]

Proveedor: OriGene
CF502972
ORIGCF502972EA 786 EUR
ORIGCF502972
Anti-PRKAR1B Mouse Monoclonal Antibody [clone: 12E5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PRKAR1B
Clonality: Monoclonal
Clone: 12E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR