Anti-TPSG1 Mouse Monoclonal Antibody [clone: 4E1]

Proveedor: OriGene
CF504162
ORIGCF504162EA 764 EUR
ORIGCF504162
Anti-TPSG1 Mouse Monoclonal Antibody [clone: 4E1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: TPSG1
Clonality: Monoclonal
Clone: 4E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR