Anti-ANAPC2 Mouse Monoclonal Antibody [clone: 1A6]

Proveedor: OriGene
ORIGCF503411EA 786 EUR
ORIGCF503411
Anti-ANAPC2 Mouse Monoclonal Antibody [clone: 1A6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ANAPC2
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR