Anti-TMPRSS5 Mouse Monoclonal Antibody [clone: 4A11]

Proveedor: OriGene
CF503409
ORIGCF503409EA 786 EUR
ORIGCF503409
Anti-TMPRSS5 Mouse Monoclonal Antibody [clone: 4A11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TMPRSS5
Clonality: Monoclonal
Clone: 4A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR